Anti-PIK3C2A Goat Antibody

Lieferant: OriGene
ORIGTA305643EA 536 EUR
ORIGTA305643
Anti-PIK3C2A Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: PIK3C2A (phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 alpha)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR