Anti-ITGAL Goat Antibody

Lieferant: OriGene
ORIGTA305639EA 547 EUR
ORIGTA305639
Anti-ITGAL Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 05-2?g/ml

Type: Primary
Antigen: ITGAL (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR