Anti-ADCYAP1 Goat Antibody

Lieferant: OriGene
ORIGTA305626EA 550 EUR
ORIGTA305626
Anti-ADCYAP1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ADCYAP1 (adenylate cyclase activating polypeptide 1 (pituitary))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR