Anti-ADORA2B Goat Antibody

Lieferant: OriGene
ORIGTA305620EA 539 EUR
ORIGTA305620
Anti-ADORA2B Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 2-4?g/ml

Type: Primary
Antigen: ADORA2B (adenosine A2b receptor)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR