Anti-FTL Goat Antibody

Lieferant: OriGene
ORIGTA305612EA 547 EUR
ORIGTA305612
Anti-FTL Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: FTL (ferritin, light polypeptide)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR