Anti-RPTOR Rabbit Monoclonal Antibody [clone: EP539Y]

Lieferant: OriGene
ORIGTA303799EA 830 EUR
ORIGTA303799
Anti-RPTOR Rabbit Monoclonal Antibody [clone: EP539Y]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunoprecipitation: 1:30; Flow Cytometry: 1:30; Immunochemistry/Immunofluorescence: 1:100-1:250

Type: Primary
Antigen: RPTOR (regulatory associated protein of MTOR, complex 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR