Anti-FOLH1 Rabbit Monoclonal Antibody [clone: EP3253]
Lieferant: OriGene
ORIGTA303519EA 800 EUR
ORIGTA303519
Anti-FOLH1 Rabbit Monoclonal Antibody [clone: EP3253]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:5000; Immunoprecipitation: 1:10; Flow Cytometry: 1:60
Type: Primary
Antigen: FOLH1 (folate hydrolase (prostate-specific membrane antigen) 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human