Anti-ESRRA Rabbit Monoclonal Antibody [clone: EPR46Y]

Lieferant: OriGene
ORIGTA303506EA 797 EUR
ORIGTA303506
Anti-ESRRA Rabbit Monoclonal Antibody [clone: EPR46Y]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Flow Cytometry: 1:20

Type: Primary
Antigen: ESRRA (estrogen-related receptor alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR