Anti-CDC25C Rabbit Monoclonal Antibody [clone: E302]
Lieferant: OriGene
ORIGTA303434EA 861 EUR
ORIGTA303434
Anti-CDC25C Rabbit Monoclonal Antibody [clone: E302]
Antikörper
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:250-1:500; Immunofluorescence: 1:250; Immunoprecipitation: 1:80
Type: Primary
Antigen: CDC25C (cell division cycle 25C)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Rat