Anti-P2RX4 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303372EA 536 EUR
ORIGTA303372
Anti-P2RX4 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: P2RX4 (purinergic receptor P2X, ligand-gated ion channel, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR