Anti-LYN Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303350EA 547 EUR
ORIGTA303350
Anti-LYN Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR