Anti-NPFFR1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303336EA 536 EUR
ORIGTA303336
Anti-NPFFR1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: NPFFR1 (neuropeptide FF receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR