Anti-UXT Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303316EA 536 EUR
ORIGTA303316
Anti-UXT Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.01-0.03?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: UXT (ubiquitously-expressed, prefoldin-like chaperone)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR