Anti-GOT1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303313EA 536 EUR
ORIGTA303313
Anti-GOT1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:4000 Western Blot: 1-3µg/ml

Type: Primary
Antigen: GOT1 (glutamic-oxaloacetic transaminase 1, soluble)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR