Anti-LASP1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303301EA 547 EUR
ORIGTA303301
Anti-LASP1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000 Western Blot: 003-01µg/ml

Type: Primary
Antigen: LASP1 (LIM and SH3 protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR