Anti-EPB41L2 Goat polyclonal antibody

Lieferant: OriGene
ORIGTA303296EA 547 EUR
ORIGTA303296
Anti-EPB41L2 Goat polyclonal antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: EPB41L2 (erythrocyte membrane protein band 4.1-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR