Anti-GALNS Goat polyclonal antibody

Lieferant: OriGene
ORIGTA303287EA 547 EUR
ORIGTA303287
Anti-GALNS Goat polyclonal antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: GALNS (galactosamine (N-acetyl)-6-sulfate sulfatase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR