Anti-CRHR1 Goat polyclonal antibody

Lieferant: OriGene
ORIGTA303282EA 536 EUR
ORIGTA303282
Anti-CRHR1 Goat polyclonal antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml; Immunofluorescence: 2?g/ml

Type: Primary
Antigen: CRHR1 (corticotropin releasing hormone receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR