Anti-BPNT1 Goat polyclonal antibody

Lieferant: OriGene
ORIGTA303272EA 547 EUR
ORIGTA303272
Anti-BPNT1 Goat polyclonal antibody
Antikörper
Recommended Dilutions: ELISA: 1:4000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: BPNT1 (3'(2'), 5'-bisphosphate nucleotidase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR