Anti-RPGRIP1L Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303270EA 536 EUR
ORIGTA303270
Anti-RPGRIP1L Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: RPGRIP1L (RPGRIP1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR