Anti-THRB Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303268EA 547 EUR
ORIGTA303268
Anti-THRB Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: THRB (thyroid hormone receptor, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR