Anti-THRA Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303267EA 547 EUR
ORIGTA303267
Anti-THRA Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000. Western Blot: 0.5-15?g/ml

Type: Primary
Antigen: THRA (thyroid hormone receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR