Anti-PINK1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303264EA 536 EUR
ORIGTA303264
Anti-PINK1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR