Anti-NRTN Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303263EA 547 EUR
ORIGTA303263
Anti-NRTN Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: NRTN (neurturin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR