Anti-PHLPP2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303258EA 547 EUR
ORIGTA303258
Anti-PHLPP2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: PHLPP2 (PH domain and leucine rich repeat protein phosphatase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR