Anti-APH1A Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303251EA 547 EUR
ORIGTA303251
Anti-APH1A Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: 005-01µg/ml

Type: Primary
Antigen: APH1A (APH1A gamma secretase subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR