Anti-PPARGC1A Goat Polyclonal Antibody

Lieferant: OriGene
TA303250
ORIGTA303250EA 547 EUR
ORIGTA303250
Anti-PPARGC1A Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: PPARGC1A (peroxisome proliferator-activated receptor gamma, coactivator 1 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR