Anti-PCK1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303224EA 547 EUR
ORIGTA303224
Anti-PCK1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: PCK1 (phosphoenolpyruvate carboxykinase 1 (soluble))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR