Anti-NACC1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303211EA 547 EUR
ORIGTA303211
Anti-NACC1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:2000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: NACC1 (nucleus accumbens associated 1, BEN and BTB (POZ) domain containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR