Anti-NANOG Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303206EA 536 EUR
ORIGTA303206
Anti-NANOG Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: 005-01µg/ml

Type: Primary
Antigen: NANOG (Nanog homeobox)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR