Anti-GPX3 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303205EA 547 EUR
ORIGTA303205
Anti-GPX3 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: GPX3 (glutathione peroxidase 3 (plasma))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR