Anti-UBASH3A Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303187EA 536 EUR
ORIGTA303187
Anti-UBASH3A Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000 Western Blot: 25µg/ml

Type: Primary
Antigen: UBASH3A (ubiquitin associated and SH3 domain containing A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR