Anti-CNTFR Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303173EA 539 EUR
ORIGTA303173
Anti-CNTFR Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: CNTFR (ciliary neurotrophic factor receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR