Anti-FOXP2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303169EA 536 EUR
ORIGTA303169
Anti-FOXP2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: FOXP2 (forkhead box P2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR