Anti-FEZ1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303166EA 539 EUR
ORIGTA303166
Anti-FEZ1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: FEZ1 (fasciculation and elongation protein zeta 1 (zygin I))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR