Anti-SMO Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303149EA 544 EUR
ORIGTA303149
Anti-SMO Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:1000 Western Blot: 01-03µg/ml

Type: Primary
Antigen: SMO (smoothened, frizzled class receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR