Anti-FBXL3 Goat polyclonal antibody

Lieferant: OriGene
ORIGTA303140EA 536 EUR
ORIGTA303140
Anti-FBXL3 Goat polyclonal antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: FBXL3 (F-box and leucine-rich repeat protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR