Anti-KCNC3 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303139EA 547 EUR
ORIGTA303139
Anti-KCNC3 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: KCNC3 (potassium voltage-gated channel, Shaw-related subfamily, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR