Anti-EBAG9 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303119EA 536 EUR
ORIGTA303119
Anti-EBAG9 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: EBAG9 (estrogen receptor binding site associated, antigen, 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR