Anti-HSP90B1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303118EA 547 EUR
ORIGTA303118
Anti-HSP90B1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: HSP90B1 (heat shock protein 90kDa beta (Grp94), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR