Anti-GPX1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303116EA 547 EUR
ORIGTA303116
Anti-GPX1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3?g/ml; Immunochemistry: 3-5?g/ml

Type: Primary
Antigen: GPX1 (glutathione peroxidase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR