Anti-AKR1C4 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303107EA 536 EUR
ORIGTA303107
Anti-AKR1C4 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:4000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: AKR1C4 (aldo-keto reductase family 1, member C4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR