Anti-STAM2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303101EA 536 EUR
ORIGTA303101
Anti-STAM2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: STAM2 (signal transducing adaptor molecule (SH3 domain and ITAM motif) 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR