Anti-LEF1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303096EA 547 EUR
ORIGTA303096
Anti-LEF1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: LEF1 (lymphoid enhancer-binding factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR