Anti-CDKN1B Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303087EA 536 EUR
ORIGTA303087
Anti-CDKN1B Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: CDKN1B (cyclin-dependent kinase inhibitor 1B (p27, Kip1))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR