Anti-STMN1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303084EA 547 EUR
ORIGTA303084
Anti-STMN1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: STMN1 (stathmin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR