Anti-DCDC2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303078EA 536 EUR
ORIGTA303078
Anti-DCDC2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:4000; Western Blot: 0.1-0.3?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: DCDC2 (doublecortin domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR