Anti-MYBL2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303072EA 536 EUR
ORIGTA303072
Anti-MYBL2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: MYBL2 (v-myb avian myeloblastosis viral oncogene homolog-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR