Anti-SUPT16H Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303070EA 547 EUR
ORIGTA303070
Anti-SUPT16H Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:8000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: SUPT16H (suppressor of Ty 16 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR