Anti-TGIF2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303055EA 536 EUR
ORIGTA303055
Anti-TGIF2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-10µg/ml

Type: Primary
Antigen: TGIF2 (TGFB-induced factor homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR